Learn More
Medline™ Multi-Layer Poly Boot Covers, Non-Skid
Catalog No. 19357611 Shop All Medline Industries Products
Encompass_Preferred
Change view
Click to view available options
Size:
Regular
X-Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-357-611 X-Large
19-357-610 Regular
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-357-611 Supplier Medline™ Supplier No. NON27143XL
This item is not returnable. View return policy
Breathable, fluid-resistant multi-layer boot covers feature secure traction for increased stability.
  • Knee-high design boot covers
  • Breathable multi-layer above the ankle and polypropylene coating below the ankle
  • Fluid protection
  • Secure traction and increased stability with special tread on the bottom
  • Heat-sealed seams help reduce risks of fluid penetration
Specifications
Size X-Large
Material SMS
Latex Free Yes
Color Blue
Sterility Non-sterile
Closure Type Elastic Top and Ankles
Sole Type Non-skid
Model NON27143XL
Seam Type Heat Sealed
Style Knee high
Includes Tread
Resistance Fluid
Product Type Boot Covers