Learn More
Enterprise Technology Solutions Medline Rubbing Alcohol, Isopropyl, 70%, 16oz, Clear
SDP
Catalog No. 5000828199
Encompass_Preferred
Rubbing Alcohol cleans minor cuts, scrapes and burns while helping to reduce the risk of getting an infection. Use small amounts of rubbing alcohol on affected areas one to three times daily. Once alcohol dries, wounds may be covered with sterile bandages. USP rubbing alcohol contains 70 percent of pure isopropyl alcohol to prevent bacteria growth.
Catalog No. 50-008-28199 Supplier Enterprise Technology Solutions Supplier No. MIIMDS098003Z
Rubbing Alcohol cleans minor cuts, scrapes and burns while helping to reduce the risk of getting an infection. Use small amounts of rubbing alcohol on affected areas one to three times daily. Once alcohol dries, wounds may be covered with sterile bandages. USP rubbing alcohol contains 70 percent of pure isopropyl alcohol to prevent bacteria growth.