Learn More
Enterprise Technology Solutions Medline Sphygmomanometer, Large, Adult Handheld, Blue
SDP
Catalog No. 5000828248
Encompass_Preferred
Large, handheld aneroid sphygmomanometer features a luminescent dial that facilitates reading over a wide range of lighting conditions. Shatter-resistant acrylic faceplate surrounds the luminescent dial. A durable zinc-alloy shell ensures protection, and the field adjustability provides exceptional long-term accuracy. No-stop pin action provides visual calibration check and added accuracy. Nylon rangefinder cuff delivers quick, convenient patient fitting. The Nite-Shift Premier handheld aneroid (PVC) is designed for adults and includes a carrying case.
Catalog No. 50-008-28248 Supplier Enterprise Technology Solutions Supplier No. MIIMDS9413
Large, handheld aneroid sphygmomanometer features a luminescent dial that facilitates reading over a wide range of lighting conditions. Shatter-resistant acrylic faceplate surrounds the luminescent dial. A durable zinc-alloy shell ensures protection, and the field adjustability provides exceptional long-term accuracy. No-stop pin action provides visual calibration check and added accuracy. Nylon rangefinder cuff delivers quick, convenient patient fitting. The Nite-Shift Premier handheld aneroid (PVC) is designed for adults and includes a carrying case.