Learn More
Enterprise Technology Solutions Medline Sphygmomanometer, PVC, Adult, Handheld, Blue
SDP
Catalog No. 5000828189
Encompass_Preferred
Handheld aneroid sphygmomanometer features a lightweight, portable and durable design. The nylon rangefinder cuff is designed to fit quickly on patients and save valuable exam time. The field adjustable aneroid also offers a no-stop pin action for visual calibration checks that ensure accuracy. Bladder, 20" tubing and bulb are made of economical PVC. Handheld aneroid is latex-free and includes a zippered carrying case.
Catalog No. 50-008-28189 Supplier Enterprise Technology Solutions Supplier No. MIIMDS9380
Handheld aneroid sphygmomanometer features a lightweight, portable and durable design. The nylon rangefinder cuff is designed to fit quickly on patients and save valuable exam time. The field adjustable aneroid also offers a no-stop pin action for visual calibration checks that ensure accuracy. Bladder, 20" tubing and bulb are made of economical PVC. Handheld aneroid is latex-free and includes a zippered carrying case.