Learn More
Enterprise Technology Solutions Medline Sterile Alcohol Prep Pads, 1"x1", 200/BX, White/Green
SDP
Catalog No. 5000828366
Encompass_Preferred
Sterile, alcohol prep pads are an easy, convenient way to cleanse the skin. These Curad Alcohol Swabs help fight bacteria at the site of application especially for pre-injection skin prepping. With 70 percent isopropyl, latex-free alcohol swabs are used most by hospitals. Two-fold, quilted swabs are individually sealed and disposable for your safety and convenience
Catalog No. 50-008-28366 Supplier Enterprise Technology Solutions Supplier No. MIICUR45581RBI
Sterile, alcohol prep pads are an easy, convenient way to cleanse the skin. These Curad Alcohol Swabs help fight bacteria at the site of application especially for pre-injection skin prepping. With 70 percent isopropyl, latex-free alcohol swabs are used most by hospitals. Two-fold, quilted swabs are individually sealed and disposable for your safety and convenience