Learn More
Enterprise Technology Solutions Medline Universal Surgical Scissors, Bandages/Clothing, 7", Black
SDP
Catalog No. 5000828197
Encompass_Preferred
Universal scissors are ideal for cutting through bandages and clothing. Sterilize easily with superheated steam under pressure of an autoclave using temperatures up to 273 degrees Fahrenheit or 133 degrees Celsius. Plastic handles are latex-free so you won't have to worry about patients with latex allergies. The handles are made of steel for durability.
Catalog No. 50-008-28197 Supplier Enterprise Technology Solutions Supplier No. MIIMDS0895018
Universal scissors are ideal for cutting through bandages and clothing. Sterilize easily with superheated steam under pressure of an autoclave using temperatures up to 273 degrees Fahrenheit or 133 degrees Celsius. Plastic handles are latex-free so you won't have to worry about patients with latex allergies. The handles are made of steel for durability.