Learn More
Enterprise Technology Solutions Medline Wound Care Kit, Hospital-Quality, 25 Pcs/BX, Assorted
SDP
Catalog No. 5000827944
Encompass_Preferred
25-piece kit wound care kit features hospital-quality essentials to help you protect, cover and cushion larger wounds after surgery or injury. Soft, absorbent, nonstick pads include adhesive tabs for quick application. Gauze pads utilize a highly absorbent cellulose layer to quickly wick away drainage. Stretch rolled gauze wraps securely around wounds without constricting to maintain a comfortable fit. Paper tape securely holds dressings in place and is excellent for delicate skin.
Catalog No. 50-008-27944 Supplier Enterprise Technology Solutions Supplier No. MIICUR1625V1
25-piece kit wound care kit features hospital-quality essentials to help you protect, cover and cushion larger wounds after surgery or injury. Soft, absorbent, nonstick pads include adhesive tabs for quick application. Gauze pads utilize a highly absorbent cellulose layer to quickly wick away drainage. Stretch rolled gauze wraps securely around wounds without constricting to maintain a comfortable fit. Paper tape securely holds dressings in place and is excellent for delicate skin.