Learn More
Medone Surgical MEDONE MICRODOSE INJECTION 5/B
SDP
Catalog No. NC2875312
Encompass
  • Injection kit designed for ophthalmic procedures.
  • Supplier is MedOne Surgical.
  • 1 mL syringe volume for precise dosing.
  • Syringe material is silicone-free to reduce contamination risk.
  • Dead-space reduction of approximately 10 µL for enhanced efficacy.
  • Removable plunger for flexibility in usage.
  • Sterilized for safety and single-use to prevent infection.
  • Pneumatic control mechanism via foot pedal for easy operation.
  • Compatible with various vitrectomy systems for versatility.
  • Luer lock connection type for secure attachment.
  • Designed for low-volume subretinal injections.
  • Kit includes connector and components for efficient use.
  • Packaged in sets of 5 per box for convenient supply.
Catalog No. NC2875312 Supplier Medone Surgical Supplier No. 3315
  • Injection kit designed for ophthalmic procedures.
  • Supplier is MedOne Surgical.
  • 1 mL syringe volume for precise dosing.
  • Syringe material is silicone-free to reduce contamination risk.
  • Dead-space reduction of approximately 10 µL for enhanced efficacy.
  • Removable plunger for flexibility in usage.
  • Sterilized for safety and single-use to prevent infection.
  • Pneumatic control mechanism via foot pedal for easy operation.
  • Compatible with various vitrectomy systems for versatility.
  • Luer lock connection type for secure attachment.
  • Designed for low-volume subretinal injections.
  • Kit includes connector and components for efficient use.
  • Packaged in sets of 5 per box for convenient supply.