Learn More
Cayman Chemical Medroxyprogesteron 100mg
SDP
Catalog No. 503015674
Encompass_Preferred
A progesterone receptor agonist (Ki = 241 nM); a metabolite of medroxyprogesterone 17-acetate; induces a conformational change in the C-terminal domain of the progesterone receptor (EC50 = 47.3 nM); increases alkaline phosphatase activity in T47D breast cancer cells (EC50 = 23 nM)
Catalog No. 50-301-5674 Supplier Cayman Chemical Supplier No. 24908100 MG
A progesterone receptor agonist (Ki = 241 nM); a metabolite of medroxyprogesterone 17-acetate; induces a conformational change in the C-terminal domain of the progesterone receptor (EC50 = 47.3 nM); increases alkaline phosphatase activity in T47D breast cancer cells (EC50 = 23 nM)