Learn More
Selleck Chemical LLC Medroxyprogesterone S3635-25mg
SDP
Catalog No. 503109752
Encompass_Preferred
Medroxyprogesterone (MP) is a synthetic pregnane steroid and a derivative of progesterone It is a potent progesterone receptor agonist
Catalog No. 50-310-9752 Supplier Selleck Chemical LLC Supplier No. S363525MG
Medroxyprogesterone (MP) is a synthetic pregnane steroid and a derivative of progesterone It is a potent progesterone receptor agonist