Learn More
Apexbio Technology LLC Medroxyprogesterone 520-85-4 50mg
SDP
Catalog No. 5000094105
Encompass_Preferred
Medroxyprogesterone (CAS 520-85-4) is a small-molecule agonist targeting the progesterone receptor It is designed to activate progesterone receptor-mediated intracellular signaling pathways thereby regulating cellular proliferation Medroxyprogesterone exerts its biological activity primarily through agonism of the progesterone receptor modulating pathways such as PI3K/Akt signaling In preclinical mouse models medroxyprogesterone demonstrates inhibitory activity on atherosclerotic lesion formation Based on these pharmacological properties medroxyprogesterone holds research potential in the study of progesterone receptor-mediated biological processes cellular proliferation and vascular pathology
Catalog No. 50-000-94105 Supplier Apexbio Technology LLC Supplier No. B347150
Medroxyprogesterone (CAS 520-85-4) is a small-molecule agonist targeting the progesterone receptor It is designed to activate progesterone receptor-mediated intracellular signaling pathways thereby regulating cellular proliferation Medroxyprogesterone exerts its biological activity primarily through agonism of the progesterone receptor modulating pathways such as PI3K/Akt signaling In preclinical mouse models medroxyprogesterone demonstrates inhibitory activity on atherosclerotic lesion formation Based on these pharmacological properties medroxyprogesterone holds research potential in the study of progesterone receptor-mediated biological processes cellular proliferation and vascular pathology