Learn More
Medchemexpress LLC Medroxyprogesterone | 520-85-4 | 98.8% | 1 G
SDP
Catalog No. 5000408444
Encompass_Preferred
Medroxyprogesterone (17α-Hydroxy-6α-methylprogesterone) is a synthetic human variant of progesterone that acts as a progesterone receptor agonist with oral activity. It can induce cell proliferation through the PI3K/Akt signaling pathway and has an inhibitory effect on atherosclerosis in mice.
  • Progesterone receptor agonist with oral activity.
  • Induces cell proliferation via PI3K/Akt signaling pathway.
  • Inhibitory effect on atherosclerosis in mice.
  • Less effective progesterone agonist activity than Medroxyprogesterone acetate.
Catalog No. 50-004-08444 Supplier Medchemexpress LLC Supplier No. HYB06481G
Medroxyprogesterone (17α-Hydroxy-6α-methylprogesterone) is a synthetic human variant of progesterone that acts as a progesterone receptor agonist with oral activity. It can induce cell proliferation through the PI3K/Akt signaling pathway and has an inhibitory effect on atherosclerosis in mice.
  • Progesterone receptor agonist with oral activity.
  • Induces cell proliferation via PI3K/Akt signaling pathway.
  • Inhibitory effect on atherosclerosis in mice.
  • Less effective progesterone agonist activity than Medroxyprogesterone acetate.