Learn More
Medchemexpress LLC Medroxyprogesterone | 520-85-4 | 98.8% | 1 ML
SDP
Catalog No. 5000408525
Encompass_Preferred
Medroxyprogesterone is a synthetic human variant of progesterone that acts as a progesterone receptor agonist with oral activity. It can induce cell proliferation through the PI3K/Akt signaling pathway and has an inhibitory effect on atherosclerosis in mice.
  • Acts as a progesterone receptor agonist with oral activity.
  • Induces cell proliferation through the PI3K/Akt signaling pathway.
  • Has an inhibitory effect on atherosclerosis in mice.
Catalog No. 50-004-08525 Supplier Medchemexpress LLC Supplier No. HYB064810MM/1ML
Medroxyprogesterone is a synthetic human variant of progesterone that acts as a progesterone receptor agonist with oral activity. It can induce cell proliferation through the PI3K/Akt signaling pathway and has an inhibitory effect on atherosclerosis in mice.
  • Acts as a progesterone receptor agonist with oral activity.
  • Induces cell proliferation through the PI3K/Akt signaling pathway.
  • Has an inhibitory effect on atherosclerosis in mice.