Learn More
Medchemexpress LLC Medroxyprogesterone | 520-85-4 | 99.3% | 50 MG
SDP
Catalog No. 5000408545
Encompass_Preferred
Medroxyprogesterone (Standard) is the analytical standard of Medroxyprogesterone, intended for research and analytical applications. This product is for research use only and not sold to patients.
  • Used as an analytical standard
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Applicable in techniques such as HPLC, GC, and MS
Catalog No. 50-004-08545 Supplier Medchemexpress LLC Supplier No. HYB0648R50MG
Medroxyprogesterone (Standard) is the analytical standard of Medroxyprogesterone, intended for research and analytical applications. This product is for research use only and not sold to patients.
  • Used as an analytical standard
  • Suitable for qualitative, quantitative, and methodological research experiments
  • Applicable in techniques such as HPLC, GC, and MS