Learn More
Apexbio Technology LLC Medroxyprogesterone acetate 71-58-9 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062359
Encompass_Preferred
Medroxyprogesterone acetate (MPA) is a synthetic steroidal derivative structurally related to the endogenous hormone progesterone classified as a progestin Acting primarily through binding to progesterone receptors it modulates gene transcription influencing endometrial proliferation and reproductive pathways Additionally MPA exhibits affinity toward glucocorticoid receptors which may contribute to its physiological effects In biomedical research MPA serves as a standard experimental tool to investigate progesterone receptor signaling mechanisms and steroid hormone interactions Furthermore it is widely used in studies focused on reproductive biology contraceptive mechanisms hormone replacement therapy models oncology research involving hormone-responsive tumors and conditions such as endometriosis
Catalog No. 50-000-62359 Supplier Apexbio Technology LLC Supplier No. B15105.1
Medroxyprogesterone acetate (MPA) is a synthetic steroidal derivative structurally related to the endogenous hormone progesterone classified as a progestin Acting primarily through binding to progesterone receptors it modulates gene transcription influencing endometrial proliferation and reproductive pathways Additionally MPA exhibits affinity toward glucocorticoid receptors which may contribute to its physiological effects In biomedical research MPA serves as a standard experimental tool to investigate progesterone receptor signaling mechanisms and steroid hormone interactions Furthermore it is widely used in studies focused on reproductive biology contraceptive mechanisms hormone replacement therapy models oncology research involving hormone-responsive tumors and conditions such as endometriosis