Learn More
Medchemexpress LLC Medroxyprogesterone acetate-d3 | 71-58-9 | 98.1% | 1 MG
SDP
Catalog No. 5000408207
Encompass_Preferred
Medroxyprogesterone acetate-d3 is a deuterium labeled version of Medroxyprogesterone acetate, a synthetic steroid known for its interaction with progesterone, androgen, and glucocorticoid receptors. This product is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08207 Supplier Medchemexpress LLC Supplier No. HYB0469S1MG
Medroxyprogesterone acetate-d3 is a deuterium labeled version of Medroxyprogesterone acetate, a synthetic steroid known for its interaction with progesterone, androgen, and glucocorticoid receptors. This product is intended for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS