Learn More
Medchemexpress LLC Medroxyprogesterone acetate-d3 | 71-58-9 | >98.1% | 5 MG
SDP
Catalog No. 5000405283
Encompass_Preferred
Medroxyprogesterone acetate-d3 is a deuterium-labeled form of Medroxyprogesterone acetate. This synthetic steroid interacts with progesterone, androgen, and glucocorticoid receptors.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable in NMR, GC-MS, or LC-MS
Catalog No. 50-004-05283 Supplier Medchemexpress LLC Supplier No. HYB0469S5MG
Medroxyprogesterone acetate-d3 is a deuterium-labeled form of Medroxyprogesterone acetate. This synthetic steroid interacts with progesterone, androgen, and glucocorticoid receptors.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis
  • Applicable in NMR, GC-MS, or LC-MS