Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Medroxyprogesterone acetate-d3 | 98.1% | 389.54 g/mol | C24H31D3O4 | 10 MG
SDP

Catalog No. 5000297808
Encompass_Preferred

Medroxyprogesterone acetate-d3 is the deuterated analog of medroxyprogesterone acetate, provided as an analytical internal standard for quantitative LC-MS and related mass spectrometry applications. It offers an isotopic match to the target analyte to improve accuracy and correct for matrix effects during sample preparation and analysis.

  • Deuterated internal standard for LC-MS quantitation.
  • Purity: 98.13% (typical, verified by HPLC).
  • Molecular weight 389.54 g/mol.
  • Chemical formula C24H31D3O4.
  • Supplied in milligram quantities suitable for analytical use.
  • Suitable for standard curve preparation and recovery experiments.

Catalog No. 50-002-97808 Supplier Medchemexpress LLC Supplier No. HYB0469S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Medroxyprogesterone acetate-d3 is the deuterated analog of medroxyprogesterone acetate, provided as an analytical internal standard for quantitative LC-MS and related mass spectrometry applications. It offers an isotopic match to the target analyte to improve accuracy and correct for matrix effects during sample preparation and analysis.

  • Deuterated internal standard for LC-MS quantitation.
  • Purity: 98.13% (typical, verified by HPLC).
  • Molecular weight 389.54 g/mol.
  • Chemical formula C24H31D3O4.
  • Supplied in milligram quantities suitable for analytical use.
  • Suitable for standard curve preparation and recovery experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.