Learn More
Medchemexpress LLC Medroxyprogesterone acetate (Standard) | 71-58-9 | 99.2% | 100 MG
SDP
Catalog No. 5000408213
Encompass_Preferred
Medroxyprogesterone acetate (Standard) is the analytical standard of Medroxyprogesterone acetate. This widely used synthetic steroid interacts with progesterone, androgen, and glucocorticoid receptors, making it suitable for various research and analytical applications.
  • Used as an analytical standard and reference standard for assay
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
Catalog No. 50-004-08213 Supplier Medchemexpress LLC Supplier No. HYB0469R100MG
Medroxyprogesterone acetate (Standard) is the analytical standard of Medroxyprogesterone acetate. This widely used synthetic steroid interacts with progesterone, androgen, and glucocorticoid receptors, making it suitable for various research and analytical applications.
  • Used as an analytical standard and reference standard for assay
  • Commonly used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications