Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Medroxyprogesterone (Standard) | 520-85-4 | 99.3% | 25 MG
SDP

Supplier:  Medchemexpress LLC 25MGHYB0648R

Encompass_Preferred

Medroxyprogesterone (Standard) is the analytical standard for Medroxyprogesterone, intended for research and analytical applications. It is a white to off-white solid with a molecular weight of 344.49 and a chemical formula of C₂₂H₃₂O₃. This product is commonly used in qualitative, quantitative, and methodological research experiments.

  • Analytical standard for Medroxyprogesterone
  • Used in qualitative, quantitative, and methodological research experiments
  • Suitable for HPLC, GC, and MS applications
  • Appears as a white to off-white solid
  • Molecular weight of 344.49
  • Chemical formula C₂₂H₃₂O₃

Catalog No. 50-004-50444


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.