Learn More
MEDTOX™ MEDTOXScan™ Cleaning Cassette
Catalog No. 22000406
Change view
Click to view available options
Intended Use:
One time use cleaning cassette to clean the internal components of the MEDTOXScan reader
Catalog No. Intended Use
22-000-406 One time use cleaning cassette to clean the internal components of the MEDTOXScan reader
1 options
Catalog No. 22-000-406 Supplier MEDTOX™ Supplier No. 833076
Add to Cart
Add to Cart
Use this Cleaning Cassette in conjunction with MEDTOXScan QC Test Devices to clean the internal components of the MEDTOXScan reader. The MEDTOXScan™ Cleaning Cassette is solely intended to clean the color Contact Imaging Sensor (CIS) in the MEDTOXScan™ Reader.
  • Cleaning Cassettes are single use, disposable devices
  • Store at room temperature (18 to 25 °C or 64 to 77°F) in unopened bag
  • Do not use past the expiration date
  • Do not use if the cassette appears damaged in any way
  • Only use mild water and mild liquid hand soap/ detergent to clean the cassette
  • Do not use an over saturated cassette in MEDTOXScan™ Reader
  • Always blot off excess water before inserting in the reader
Specifications
Intended Use One time use cleaning cassette to clean the internal components of the MEDTOXScan reader