Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEDTOX™ MEDTOXScan™ Printer Paper Roll

Catalog No. 21500132
Change view
Click to view available options
Quantity:
Roll or Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-500-132 Roll or Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-500-132 Supplier MEDTOX™ Supplier No. 101885
Add to Cart
Add to Cart

MEDTOX Diagnostics, Inc.™ MEDTOXScan™ Printer Paper Roll is used in the printer that prints out test results from MEDTOXScan Reader.

  • Paper for use with MEDTOXScan reader
  • Order as roll or individually

Specifications

Content And Storage Ambient
For Use With (Equipment) MEDTOXScan Reader
Type Printer Paper
Quantity Roll or Each
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.