Learn More
ATCC MEF (CF-1) IRR; Mouse fibroblast
Supplier:  ATCC SCRC1040.1
Encompass
MEF (CF-1) IRR is a fibroblast cell that was isolated from an embryo of a mouse. The cells can be used as irradiated cells for use as a feeder layer.
Catalog No. 50-238-2112
 Restricted Item