Learn More
ATCC MEF (CF-1) MITC; Mouse fibroblast
Catalog No. 502382725
Encompass
MEF (CF-1) MITC is a fibroblast cell that was isolated from an embryo of a mouse. The cells can be used as a feeder layer.
Catalog No. 50-238-2725 Supplier ATCC Supplier No. SCRC1040.2A
MEF (CF-1) MITC is a fibroblast cell that was isolated from an embryo of a mouse. The cells can be used as a feeder layer.