Learn More
ATCC MEF (CF-1); Mouse fibroblast
Supplier:  ATCC SCRC1040
Encompass
MEF (CF-1) is a fibroblast cell that was isolated from an embryo of a mouse. The cells can be used as a feeder layer to support the growth of embryonic stem (ES) cells and for the maintenance of ES cells in the undifferentiated state.
Catalog No. 50-238-3371