Learn More
Lonza Walkersville Mouse Embryonic Fibroblast Nucleofector Kit 2
Catalog No. NC9106405
Encompass
The Nucleofector 2b Device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. The Mouse Embryonic Fibroblast Nucleofector Kit 2 is one of our two kits suited for efficient transfectiion of primary mouse embyonic fibroblasts (MEF). Due to significant diversity between different MEF clones, we have developed two Nucleofector Kits (1 and 2) and an easy-to-use Nucleofector Starter Kit to determine which of the 2 kits is optimal for a specific MEF clone.Benefits:High transfection efficiencies for primary cellsCells maintain functionality post transfectionApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cells -Low-throughput transfection in single cuvette formatContent:2.25 mL Nucleofector Solution for Mouse Embryonic Fibroblasts0.5 mL Supplement 130 µg pmaxGFP Vector (0.5 μg/μl in 10 mM Tris pH 8.0)25 Aluminum cuvette (100 µL)25 Single use pipettes
Catalog No. NC9106405 Supplier Lonza Walkersville Supplier No. VPD1005
The Nucleofector 2b Device (or older generations I or II) works cell type specific kits, each of them dedicated to an individual primary cell. The Mouse Embryonic Fibroblast Nucleofector Kit 2 is one of our two kits suited for efficient transfectiion of primary mouse embyonic fibroblasts (MEF). Due to significant diversity between different MEF clones, we have developed two Nucleofector Kits (1 and 2) and an easy-to-use Nucleofector Starter Kit to determine which of the 2 kits is optimal for a specific MEF clone.Benefits:High transfection efficiencies for primary cellsCells maintain functionality post transfectionApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cells -Low-throughput transfection in single cuvette formatContent:2.25 mL Nucleofector Solution for Mouse Embryonic Fibroblasts0.5 mL Supplement 130 µg pmaxGFP Vector (0.5 μg/μl in 10 mM Tris pH 8.0)25 Aluminum cuvette (100 µL)25 Single use pipettes