Learn More
Aviva Systems Biology Corp MEF2D Antibody - middle region (ARP38699_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP38699_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MEF2D. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MEF2D. Alternate names: DKFZp686I1536ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-62935