Learn More
Medchemexpress LLC HY-B0574 500mg Medchemexpress, Mefenamic acid CAS:61-68-7 Purity:>98%
SDP
Catalog No. 502082606
Encompass_Preferred
Medchemexpress, HY-B0574 500mg Mefenamic acid CAS:61-68-7 Mefenamic acid is a non-steroidal anti-inflammatory agent, acting as a competitive inhibitor of hCOX-1 and hCOX-2, with IC50s of 40 nM and 3 μM for hCOX-1 and hCOX-2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2606 Supplier Medchemexpress LLC Supplier No. HYB0574500MG
Medchemexpress, HY-B0574 500mg Mefenamic acid CAS:61-68-7 Mefenamic acid is a non-steroidal anti-inflammatory agent, acting as a competitive inhibitor of hCOX-1 and hCOX-2, with IC50s of 40 nM and 3 μM for hCOX-1 and hCOX-2, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.