Learn More
Cayman Chemical MefenamIc AcId 500mg
SDP
Catalog No. 502964487
Encompass_Preferred
A NSAID and COX-2 inhibitor; binds to COX-2 (Kd = 4 nM) and inhibits COX-2-dependent oxygenation of arachidonic acid in vitro (Ki = 10 µM); reduces acetic acid-induced writhing in rats at 30 mg/kg; inhibits DNBC-induced increases in skin thickness in a mouse model of delayed-type hypersensitivity; reduces the antibody response to sheep red blood cells in mice
Catalog No. 50-296-4487 Supplier Cayman Chemical Supplier No. 23650500 MG
A NSAID and COX-2 inhibitor; binds to COX-2 (Kd = 4 nM) and inhibits COX-2-dependent oxygenation of arachidonic acid in vitro (Ki = 10 µM); reduces acetic acid-induced writhing in rats at 30 mg/kg; inhibits DNBC-induced increases in skin thickness in a mouse model of delayed-type hypersensitivity; reduces the antibody response to sheep red blood cells in mice