Learn More
Apexbio Technology LLC Mefenamic Acid 61-68-7 5g
SDP
Catalog No. 5000091363
Encompass_Preferred
Mefenamic acid (CAS 61-68-7) is a non-steroidal anti-inflammatory compound that acts by inhibiting cyclooxygenase (COX) enzymes leading to decreased synthesis of prostaglandins involved in inflammation and pain signaling It is extensively studied for its ability to modulate inflammatory pathways and is commonly utilized as a pharmacological tool in research focused on pain mechanisms inflammatory responses and prostaglandin-mediated processes Mefenamic acid is often employed in in vitro and in vivo models to evaluate the effects of COX inhibition on physiological and pathological conditions
Catalog No. 50-000-91363 Supplier Apexbio Technology LLC Supplier No. B14495000
Add to Cart
Add to Cart
Mefenamic acid (CAS 61-68-7) is a non-steroidal anti-inflammatory compound that acts by inhibiting cyclooxygenase (COX) enzymes leading to decreased synthesis of prostaglandins involved in inflammation and pain signaling It is extensively studied for its ability to modulate inflammatory pathways and is commonly utilized as a pharmacological tool in research focused on pain mechanisms inflammatory responses and prostaglandin-mediated processes Mefenamic acid is often employed in in vitro and in vivo models to evaluate the effects of COX inhibition on physiological and pathological conditions