Learn More
Medchemexpress LLC Mefenamic Acid-d3 | 1189707-81-0 | 2.5 MG
SDP
Catalog No. 5000408669
Encompass_Preferred
Mefenamic Acid-d3 is the deuterium labeled Mefenamic acid. Mefenamic acid is a BBB-permeable non-steroidal anti-inflammatory agent, acting as a competitive inhibitor of hCOX-1 and hCOX-2, with IC50s of 40 nM and 3 μM, respectively. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled mefenamic acid
  • BBB-permeable non-steroidal anti-inflammatory agent
  • Acts as a competitive inhibitor of hCOX-1 and hCOX-2
  • Suitable as a tracer for research
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-08669 Supplier Medchemexpress LLC Supplier No. HYB0574S12.5MG
Mefenamic Acid-d3 is the deuterium labeled Mefenamic acid. Mefenamic acid is a BBB-permeable non-steroidal anti-inflammatory agent, acting as a competitive inhibitor of hCOX-1 and hCOX-2, with IC50s of 40 nM and 3 μM, respectively. It can be used as a tracer or an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuterium labeled mefenamic acid
  • BBB-permeable non-steroidal anti-inflammatory agent
  • Acts as a competitive inhibitor of hCOX-1 and hCOX-2
  • Suitable as a tracer for research
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS