Learn More
Medchemexpress LLC Mefenamic acid-d4 | 1216745-79-7 | MFCD08063563 | 98.0% | 5 MG
SDP
Catalog No. 5000408532
Encompass_Preferred
Mefenamic acid-d4 is a deuterium-labeled form of Mefenamic acid, a BBB-permeable non-steroidal anti-inflammatory agent. It functions as a competitive inhibitor of both hCOX-1 and hCOX-2, with respective IC50 values of 40 nM and 3 μM. This compound is intended for research applications only and is not suitable for patient use.
  • Can be used as a tracer in various research applications.
  • Suitable for use as an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS.
  • Acts as an inhibitor of both hCOX-1 and hCOX-2.
  • Appears as a solid, white to off-white substance.
  • Exhibits solubility in DMSO (≥ 100 mg/mL) and limited solubility in H2O (0.1 mg/mL).
  • Recommended storage for powder is -20°C for up to 3 years; in solvent, store at -80°C for 6 months or -20°C for 1 month.
Catalog No. 50-004-08532 Supplier Medchemexpress LLC Supplier No. HYB0574S5MG
Mefenamic acid-d4 is a deuterium-labeled form of Mefenamic acid, a BBB-permeable non-steroidal anti-inflammatory agent. It functions as a competitive inhibitor of both hCOX-1 and hCOX-2, with respective IC50 values of 40 nM and 3 μM. This compound is intended for research applications only and is not suitable for patient use.
  • Can be used as a tracer in various research applications.
  • Suitable for use as an internal standard for quantitative analysis through techniques like NMR, GC-MS, or LC-MS.
  • Acts as an inhibitor of both hCOX-1 and hCOX-2.
  • Appears as a solid, white to off-white substance.
  • Exhibits solubility in DMSO (≥ 100 mg/mL) and limited solubility in H2O (0.1 mg/mL).
  • Recommended storage for powder is -20°C for up to 3 years; in solvent, store at -80°C for 6 months or -20°C for 1 month.