Learn More
Medchemexpress LLC Mefenpyr-diethyl | 135590-91-9 | 99.5% | 373.23 | 1 G
Supplier: Medchemexpress LLC HY1363761G
Mefenpyr-diethyl is a pyrazoline-type herbicide safener that enhances lipid biosynthesis. It effectively prevents herbicide-induced phytotoxic damage caused by herbicides such as fenoxaprop-p-ethyl and mesosulfuron-methyl, protecting cereal crops including wheat. Mefenpyr-diethyl significantly increases lipid content in wheat when applied alone or in combination with fenoxaprop-p-ethyl. Its mechanism of action does not involve the induction of glutathione S-transferase (GST) activity in wheat.
- Functions as a pyrazoline-type herbicide safener
- Enhances lipid biosynthesis
- Effectively prevents herbicide-induced phytotoxic damage
- Protects cereal crops, including wheat
- Significantly increases lipid content in wheat
- Mechanism of action does not involve induction of glutathione S-transferase activity