Learn More
Medchemexpress LLC Mefenpyr-diethyl | 135590-91-9 | 99.5% | 373.23 | 10 G
SDP
Catalog No. 5000350274
Encompass_Preferred
Mefenpyr-diethyl is a pyrazoline-type herbicide safener that functions to enhance lipid biosynthesis. It effectively prevents herbicide-induced phytotoxic damage caused by herbicides like fenoxaprop-p-ethyl and mesosulfuron-methyl, thereby protecting cereal crops, including wheat. It significantly increases lipid content in wheat, whether applied alone or in combination with fenoxaprop-p-ethyl. Its mechanism of action does not involve inducing glutathione S-transferase (GST) activity in wheat.
  • Functions as a pyrazoline-type herbicide safener
  • Enhances lipid biosynthesis
  • Prevents herbicide-induced phytotoxic damage in cereal crops
  • Protects against damage from herbicides like fenoxaprop-p-ethyl and mesosulfuron-methyl
  • Significantly increases lipid content in wheat
  • Does not involve inducing glutathione S-transferase (GST) activity in wheat
  • Alters glutathione S-transferase (GST) activity in wheat cultivars
Catalog No. 50-003-50274 Supplier Medchemexpress LLC Supplier No. HY13637610G
Mefenpyr-diethyl is a pyrazoline-type herbicide safener that functions to enhance lipid biosynthesis. It effectively prevents herbicide-induced phytotoxic damage caused by herbicides like fenoxaprop-p-ethyl and mesosulfuron-methyl, thereby protecting cereal crops, including wheat. It significantly increases lipid content in wheat, whether applied alone or in combination with fenoxaprop-p-ethyl. Its mechanism of action does not involve inducing glutathione S-transferase (GST) activity in wheat.
  • Functions as a pyrazoline-type herbicide safener
  • Enhances lipid biosynthesis
  • Prevents herbicide-induced phytotoxic damage in cereal crops
  • Protects against damage from herbicides like fenoxaprop-p-ethyl and mesosulfuron-methyl
  • Significantly increases lipid content in wheat
  • Does not involve inducing glutathione S-transferase (GST) activity in wheat
  • Alters glutathione S-transferase (GST) activity in wheat cultivars