Learn More
Medchemexpress LLC Mefenpyr-diethyl | 135590-91-9 | 99.5% | 373.23 | 10 MM 1 ML
SDP
Catalog No. 5000351596
Encompass_Preferred
Mefenpyr-diethyl is a pyrazoline-type herbicide safener that enhances lipid biosynthesis. It prevents herbicide-induced phytotoxic damage caused by herbicides in cereal crops like wheat. This compound significantly increases lipid content in wheat when applied alone or with fenoxaprop-p-ethyl. Its mechanism of action does not involve inducing glutathione S-transferase activity in wheat.
  • Enhances lipid biosynthesis
  • Prevents herbicide-induced phytotoxic damage from herbicides such as fenoxaprop-p-ethyl and mesosulfuron-methyl
  • Protects cereal crops like wheat
  • Significantly increases lipid content in wheat whether applied alone or with fenoxaprop-p-ethyl
  • Does not involve inducing glutathione S-transferase activity in wheat
  • Prevents phytotoxicity in wheat cultivars BRS 49, CD 104, CEP 24, IAPAR 78, and Rubi
Catalog No. 50-003-51596 Supplier Medchemexpress LLC Supplier No. HY13637610MM/1ML
Mefenpyr-diethyl is a pyrazoline-type herbicide safener that enhances lipid biosynthesis. It prevents herbicide-induced phytotoxic damage caused by herbicides in cereal crops like wheat. This compound significantly increases lipid content in wheat when applied alone or with fenoxaprop-p-ethyl. Its mechanism of action does not involve inducing glutathione S-transferase activity in wheat.
  • Enhances lipid biosynthesis
  • Prevents herbicide-induced phytotoxic damage from herbicides such as fenoxaprop-p-ethyl and mesosulfuron-methyl
  • Protects cereal crops like wheat
  • Significantly increases lipid content in wheat whether applied alone or with fenoxaprop-p-ethyl
  • Does not involve inducing glutathione S-transferase activity in wheat
  • Prevents phytotoxicity in wheat cultivars BRS 49, CD 104, CEP 24, IAPAR 78, and Rubi