Learn More
Medchemexpress LLC Mefenpyr-diethyl | 135590-91-9 | MFCD09753375 | 99.5% | 373.23 | 2 G
SDP
Catalog No. 5000349458
Encompass_Preferred
Mefenpyr-diethyl is a pyrazoline-type herbicide safener that functions by enhancing lipid biosynthesis. It effectively prevents herbicide-induced phytotoxic damage caused by certain herbicides, protecting cereal crops such as wheat. This compound significantly increases lipid content in wheat, whether applied alone or in combination with fenoxaprop-p-ethyl. Its mechanism of action does not involve the induction of glutathione S-transferase (GST) activity in wheat.
  • Prevents herbicide-induced phytotoxicity in wheat cultivars.
  • Enhances lipid biosynthesis in cereal crops.
  • Effective when applied alone or with fenoxaprop-p-ethyl.
  • Solid appearance, off-white to light yellow color.
  • Store at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.
Catalog No. 50-003-49458 Supplier Medchemexpress LLC Supplier No. HY1363762G
Mefenpyr-diethyl is a pyrazoline-type herbicide safener that functions by enhancing lipid biosynthesis. It effectively prevents herbicide-induced phytotoxic damage caused by certain herbicides, protecting cereal crops such as wheat. This compound significantly increases lipid content in wheat, whether applied alone or in combination with fenoxaprop-p-ethyl. Its mechanism of action does not involve the induction of glutathione S-transferase (GST) activity in wheat.
  • Prevents herbicide-induced phytotoxicity in wheat cultivars.
  • Enhances lipid biosynthesis in cereal crops.
  • Effective when applied alone or with fenoxaprop-p-ethyl.
  • Solid appearance, off-white to light yellow color.
  • Store at 4°C, protected from light.
  • When in solvent, store at -80°C for 6 months or -20°C for 1 month, protected from light.