Learn More
Medchemexpress LLC HY-136063 5mg Medchemexpress, Mefentrifluconazole CAS:1417782-03-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1360635MG
Encompass_Preferred
Medchemexpress, HY-136063 5mg Mefentrifluconazole CAS:1417782-03-6 Mefentrifluconazole is a novel azole derivative and used as an agrochemical broad-spectrum antifungal agent. Mefentrifluconazole is a potent, selective and orally active fungal CYP51 (Kd= 0.5 nM) inhibitor, but shows less inhibitory activity on human aromatase (IC50=0.92 μM). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0392