Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mefexamide hydrochloride | 3413-64-7 | 99.5% | 100 MG
SDP

Catalog No. 5000408834
Encompass_Preferred

Mefexamide hydrochloride is a psychostimulant capable of reversing the psychodepressant effects induced by diazepam and benzodiazepine sedatives. This product is intended for research use only.

  • Reverses psychodepressant effects
  • Purity of 99.45%
  • Solid appearance, white to off-white color
  • Recommended storage at 4°C, sealed, and away from moisture
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C

Catalog No. 50-004-08834 Supplier Medchemexpress LLC Supplier No. HYB0950A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mefexamide hydrochloride is a psychostimulant capable of reversing the psychodepressant effects induced by diazepam and benzodiazepine sedatives. This product is intended for research use only.

  • Reverses psychodepressant effects
  • Purity of 99.45%
  • Solid appearance, white to off-white color
  • Recommended storage at 4°C, sealed, and away from moisture
  • In solvent, stable for 6 months at -80°C or 1 month at -20°C

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.