Learn More
TARGETMOL CHEMICALS INC Mefloquine 1MG
SDP
Catalog No. 502880637
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Mefloquine (Ro 215998), a quinoline antimalarial agent, is an anti-SARS-CoV-2 entry inhibitor. Mefloquine is also a K+ channel (KvQT1/minK) antagonist with an IC50 of ~1 uM. Mefloquine can be used for malaria, systemic lupus erythematosus, and cancer research. Purity 99.97%
Catalog No. 50-288-0637 Supplier TARGETMOL CHEMICALS INC Supplier No. T0860L1MG
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Mefloquine (Ro 215998), a quinoline antimalarial agent, is an anti-SARS-CoV-2 entry inhibitor. Mefloquine is also a K+ channel (KvQT1/minK) antagonist with an IC50 of ~1 uM. Mefloquine can be used for malaria, systemic lupus erythematosus, and cancer research. Purity 99.97%