Learn More
Medchemexpress LLC Mefloquine | 53230-10-7 | 100.0% | 378.31 | 5 MG
SDP
Catalog No. 5000386613
Encompass_Preferred
Mefloquine is an orally active and potent quinoline antimalarial agent, also functioning as an anti-SARS-CoV-2 entry inhibitor and a K+ channel (KvQT1/minK) antagonist. It is suitable for various research applications.
  • Orally active and potent quinoline antimalarial agent.
  • Anti-SARS-CoV-2 entry inhibitor.
  • K+ channel (KvQT1/minK) antagonist with an IC50 of approximately 1 μM.
  • Suitable for malaria research.
  • Suitable for systemic lupus erythematosus research.
  • Suitable for cancer research.
Catalog No. 50-003-86613 Supplier Medchemexpress LLC Supplier No. HY174375MG
Mefloquine is an orally active and potent quinoline antimalarial agent, also functioning as an anti-SARS-CoV-2 entry inhibitor and a K+ channel (KvQT1/minK) antagonist. It is suitable for various research applications.
  • Orally active and potent quinoline antimalarial agent.
  • Anti-SARS-CoV-2 entry inhibitor.
  • K+ channel (KvQT1/minK) antagonist with an IC50 of approximately 1 μM.
  • Suitable for malaria research.
  • Suitable for systemic lupus erythematosus research.
  • Suitable for cancer research.