Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mefloquine | 53230-10-7 | 100.0% | 378.31 g·mol-1 | C17H16F6N2O | 10 MG
SDP

Catalog No. 5000299130
Encompass_Preferred

Mefloquine is an orally active quinoline antimalarial agent used in biomedical research. It has reported activity as an inhibitor of SARS-CoV-2 entry and as a K+ channel antagonist (IC50 ≈1 μM). Supplied as a solid for research use only; not intended for clinical or human therapeutic use.

  • High purity (HPLC 99.96%).
  • Chemical formula C17H16F6N2O; molecular weight 378.31 g·mol-1.
  • CAS number 53230-10-7.
  • Common uses: malaria, autoimmune disease, and oncology research.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-99130 Supplier Medchemexpress LLC Supplier No. HY1743710MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mefloquine is an orally active quinoline antimalarial agent used in biomedical research. It has reported activity as an inhibitor of SARS-CoV-2 entry and as a K+ channel antagonist (IC50 ≈1 μM). Supplied as a solid for research use only; not intended for clinical or human therapeutic use.

  • High purity (HPLC 99.96%).
  • Chemical formula C17H16F6N2O; molecular weight 378.31 g·mol-1.
  • CAS number 53230-10-7.
  • Common uses: malaria, autoimmune disease, and oncology research.
  • Storage: powder -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.