Learn More
Medchemexpress LLC Mefloquine | 53230-10-7 | 99.96% | 378.31 | 200 MG
SDP
Catalog No. 5000386189
Encompass_Preferred
Mefloquine is an orally active and potent quinoline antimalarial agent that also functions as an anti-SARS-CoV-2 entry inhibitor. It is a K+ channel (KvQT1/minK) antagonist with an IC50 of approximately 1 μM. It can be utilized for research related to malaria, systemic lupus erythematosus, and cancer, selectively inhibiting prostate cancer (PCa) cell growth with an IC50 of ~10 μM by inducing hyperpolarization of mitochondrial membrane potential and ROS generation.
  • Orally active and potent quinoline antimalarial agent
  • Functions as an anti-SARS-CoV-2 entry inhibitor
  • K+ channel (KvQT1/minK) antagonist with an IC50 of approximately 1 μM
  • Can be used for malaria, systemic lupus erythematosus, and cancer research
  • Selectively inhibits prostate cancer cell growth
Catalog No. 50-003-86189 Supplier Medchemexpress LLC Supplier No. HY17437200MG
Mefloquine is an orally active and potent quinoline antimalarial agent that also functions as an anti-SARS-CoV-2 entry inhibitor. It is a K+ channel (KvQT1/minK) antagonist with an IC50 of approximately 1 μM. It can be utilized for research related to malaria, systemic lupus erythematosus, and cancer, selectively inhibiting prostate cancer (PCa) cell growth with an IC50 of ~10 μM by inducing hyperpolarization of mitochondrial membrane potential and ROS generation.
  • Orally active and potent quinoline antimalarial agent
  • Functions as an anti-SARS-CoV-2 entry inhibitor
  • K+ channel (KvQT1/minK) antagonist with an IC50 of approximately 1 μM
  • Can be used for malaria, systemic lupus erythematosus, and cancer research
  • Selectively inhibits prostate cancer cell growth