Learn More
Medchemexpress LLC Mefloquine | 53230-10-7 | 99.96% | 378.31 | 50 MG
SDP
Catalog No. 5000386247
Encompass_Preferred
Mefloquine (Standard) is the analytical standard of Mefloquine, intended for research and analytical applications. It is an orally active and potent quinoline antimalarial agent and an anti-SARS-CoV-2 entry inhibitor. Mefloquine also acts as a K+ channel (KvQT1/minK) antagonist with an IC50 of approximately 1 μM. It can be utilized for malaria, systemic lupus erythematosus, and cancer research.
  • Analytical standard of Mefloquine.
  • Intended for research and analytical applications.
  • Orally active and potent quinoline antimalarial agent.
  • Anti-SARS-CoV-2 entry inhibitor.
  • K+ channel (KvQT1/minK) antagonist with an IC50 of ~1 μM.
  • Applicable for malaria, systemic lupus erythematosus, and cancer research.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
Catalog No. 50-003-86247 Supplier Medchemexpress LLC Supplier No. HY1743750MG
Mefloquine (Standard) is the analytical standard of Mefloquine, intended for research and analytical applications. It is an orally active and potent quinoline antimalarial agent and an anti-SARS-CoV-2 entry inhibitor. Mefloquine also acts as a K+ channel (KvQT1/minK) antagonist with an IC50 of approximately 1 μM. It can be utilized for malaria, systemic lupus erythematosus, and cancer research.
  • Analytical standard of Mefloquine.
  • Intended for research and analytical applications.
  • Orally active and potent quinoline antimalarial agent.
  • Anti-SARS-CoV-2 entry inhibitor.
  • K+ channel (KvQT1/minK) antagonist with an IC50 of ~1 μM.
  • Applicable for malaria, systemic lupus erythematosus, and cancer research.
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.