Learn More
Selleck Chemical LLC Mefloquine HCl S4420-100mg
SDP
Catalog No. 503108132
Encompass_Preferred
Mefloquine HCl(Mefloquin hydrochloride) is a blood schizonticide by inhibiting hemozoin formation used as an antimalarial drug
Catalog No. 50-310-8132 Supplier Selleck Chemical LLC Supplier No. S4420100MG
Mefloquine HCl(Mefloquin hydrochloride) is a blood schizonticide by inhibiting hemozoin formation used as an antimalarial drug