Learn More
Selleck Chemical LLC Mefloquine HCl S4420-500mg
SDP
Catalog No. 503110249
Encompass_Preferred
Mefloquine HCl(Mefloquin hydrochloride) is a blood schizonticide by inhibiting hemozoin formation used as an antimalarial drug
Catalog No. 50-311-0249 Supplier Selleck Chemical LLC Supplier No. S4420500MG
Mefloquine HCl(Mefloquin hydrochloride) is a blood schizonticide by inhibiting hemozoin formation used as an antimalarial drug