Learn More
Medchemexpress LLC Mefloquine (Standard) | 53230-10-7 | 378.31 | 1 G
SDP
Catalog No. 5000386416
Encompass_Preferred
Mefloquine (Standard) is the analytical standard of Mefloquine, intended for research and analytical applications. It is an orally active and potent quinoline antimalarial agent, and also functions as an anti-SARS-CoV-2 entry inhibitor. Mefloquine is a K+ channel (KvQT1/minK) antagonist with an IC50 of ~1 μM. It can be utilized for malaria, systemic lupus erythematosus, and cancer research.
  • Analytical standard grade
  • Reference standard supplied assay
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Suitable for various research and analytical applications
  • Potential for malaria, systemic lupus erythematosus, and cancer research
Catalog No. 50-003-86416 Supplier Medchemexpress LLC Supplier No. HY174371G
Mefloquine (Standard) is the analytical standard of Mefloquine, intended for research and analytical applications. It is an orally active and potent quinoline antimalarial agent, and also functions as an anti-SARS-CoV-2 entry inhibitor. Mefloquine is a K+ channel (KvQT1/minK) antagonist with an IC50 of ~1 μM. It can be utilized for malaria, systemic lupus erythematosus, and cancer research.
  • Analytical standard grade
  • Reference standard supplied assay
  • Used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS
  • Suitable for various research and analytical applications
  • Potential for malaria, systemic lupus erythematosus, and cancer research