Learn More
Medchemexpress LLC Mefloquine (Standard) | 53230-10-7 | MFCD00869324 | 378.31 | 100 MG
SDP
Catalog No. 5000386412
Encompass_Preferred
Mefloquine (Standard) is the analytical standard of Mefloquine, intended for research and analytical applications. It is an orally active and potent quinoline antimalarial agent and an anti-SARS-CoV-2 entry inhibitor. Mefloquine is also a K+ channel (KvQT1/minK) antagonist with an IC50 of ~1 μM. This analytical standard grade compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Orally active and potent quinoline antimalarial agent.
  • Anti-SARS-CoV-2 entry inhibitor.
  • K+ channel (KvQT1/minK) antagonist.
  • Used for malaria, systemic lupus erythematosus, and cancer research.
  • Analytical standard grade for qualitative, quantitative, and methodological research experiments.
Catalog No. 50-003-86412 Supplier Medchemexpress LLC Supplier No. HY17437100MG
Mefloquine (Standard) is the analytical standard of Mefloquine, intended for research and analytical applications. It is an orally active and potent quinoline antimalarial agent and an anti-SARS-CoV-2 entry inhibitor. Mefloquine is also a K+ channel (KvQT1/minK) antagonist with an IC50 of ~1 μM. This analytical standard grade compound is commonly used in qualitative, quantitative, and methodological research experiments in HPLC, GC, and MS.
  • Orally active and potent quinoline antimalarial agent.
  • Anti-SARS-CoV-2 entry inhibitor.
  • K+ channel (KvQT1/minK) antagonist.
  • Used for malaria, systemic lupus erythematosus, and cancer research.
  • Analytical standard grade for qualitative, quantitative, and methodological research experiments.