Learn More
AdipoGen Mefluidide
SDP
Supplier:  AdipoGen CDXM0134M050
Encompass_Preferred
Chemical. CAS 53780-34-0. Formula C11H13F3N2O3S. MW 310.3. Synthetic. Trifluoromethanesulphonanilide herbicide. Plant growth regulator based on the inhibition of distinct 3-ketoacyl-CoA synthase KCS isoenzymes in very-long-chain fatty acid VLCFA synthesis. Dependent on the affected KCS isoenzymes and the intensity of inhibition, plant phenotypes are induced which are characterized by shoot stunting with intensified green pigmentation or strong growth inhibition followed by tissue necrosis and plant death. Compound can be used as analytical reference material.
Catalog No. 50-256-0502