Learn More
Medchemexpress LLC Mefuparib hydrochloride | 1449746-00-2 | 98.6% | 334.77 | 5 MG
SDP
Catalog No. 5000364597
Encompass_Preferred
Mefuparib hydrochloride is an orally active, substrate-competitive, and selective inhibitor of PARP1/2, with IC50 values of 3.2 nM and 1.9 nM respectively. It is known to induce apoptosis and exhibits significant anticancer activity both in vitro and in vivo.
  • Orally active.
  • Substrate-competitive and selective PARP1/2 inhibitor.
  • Induces apoptosis.
  • Possesses prominent anticancer activity in vitro and in vivo.
Catalog No. 50-003-64597 Supplier Medchemexpress LLC Supplier No. 5MGHY122661
Mefuparib hydrochloride is an orally active, substrate-competitive, and selective inhibitor of PARP1/2, with IC50 values of 3.2 nM and 1.9 nM respectively. It is known to induce apoptosis and exhibits significant anticancer activity both in vitro and in vivo.
  • Orally active.
  • Substrate-competitive and selective PARP1/2 inhibitor.
  • Induces apoptosis.
  • Possesses prominent anticancer activity in vitro and in vivo.