Learn More
Aviva Systems Biology Corp MEFV Antibody - N-terminal region (ARP32016_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP32016_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against MEFV. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human MEFV. Alternate names: FMF, MEF, TRIM20ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54424